Learn More
Medchemexpress LLC 4-Hydroxyindole | 2380-94-1 | 133.15 | 1 ML
SDP
Catalog No. 5000405048
Encompass_Preferred
4-Hydroxyindole is a type of hydroxyindole where the 1H-indole at position 4 is substituted by a hydroxyl group. It serves as an important raw material or intermediate in the synthesis of pharmaceutical products and industrial polymers. 4-Hydroxyindole inhibits amyloid fibrillization, induces liver function impairment, thyroid abnormalities, and blood glucose fluctuations in mice. This compound holds potential for research in neurodegenerative diseases and metabolic disorders.
  • Serves as an important raw material or intermediate in the synthesis of pharmaceutical products and industrial polymers.
  • Inhibits amyloid fibrillization.
  • Holds potential for research in neurodegenerative diseases and metabolic disorders.
Catalog No. 50-004-05048 Supplier Medchemexpress LLC Supplier No. HY3459610MM/1ML
4-Hydroxyindole is a type of hydroxyindole where the 1H-indole at position 4 is substituted by a hydroxyl group. It serves as an important raw material or intermediate in the synthesis of pharmaceutical products and industrial polymers. 4-Hydroxyindole inhibits amyloid fibrillization, induces liver function impairment, thyroid abnormalities, and blood glucose fluctuations in mice. This compound holds potential for research in neurodegenerative diseases and metabolic disorders.
  • Serves as an important raw material or intermediate in the synthesis of pharmaceutical products and industrial polymers.
  • Inhibits amyloid fibrillization.
  • Holds potential for research in neurodegenerative diseases and metabolic disorders.