Learn More
Medchemexpress LLC 4-Hydroxyindole | 2380-94-1 | 133.15 | 25 MG
SDP
Catalog No. 5000402726
Encompass_Preferred
4-Hydroxyindole is a type of hydroxyindole where the 1H-indole at position 4 is substituted by a hydroxyl group. It is an important raw material or intermediate in the synthesis of pharmaceutical products and industrial polymers. This compound inhibits amyloid fibrillization and induces liver function impairment, thyroid abnormalities, and blood glucose fluctuations in mice, holding potential for research in neurodegenerative diseases and metabolic disorders.
  • Important raw material or intermediate in the synthesis of pharmaceutical products and industrial polymers.
  • Inhibits amyloid fibrillization.
  • Induces liver function impairment, thyroid abnormalities, and blood glucose fluctuations in mice.
  • Holds potential for research in neurodegenerative diseases and metabolic disorders.
Catalog No. 50-004-02726 Supplier Medchemexpress LLC Supplier No. 25MGHY34596R
4-Hydroxyindole is a type of hydroxyindole where the 1H-indole at position 4 is substituted by a hydroxyl group. It is an important raw material or intermediate in the synthesis of pharmaceutical products and industrial polymers. This compound inhibits amyloid fibrillization and induces liver function impairment, thyroid abnormalities, and blood glucose fluctuations in mice, holding potential for research in neurodegenerative diseases and metabolic disorders.
  • Important raw material or intermediate in the synthesis of pharmaceutical products and industrial polymers.
  • Inhibits amyloid fibrillization.
  • Induces liver function impairment, thyroid abnormalities, and blood glucose fluctuations in mice.
  • Holds potential for research in neurodegenerative diseases and metabolic disorders.