Learn More
Medchemexpress LLC 4-Hydroxyindole | 2380-94-1 | 133.15 | 5 G
SDP
Catalog No. 5000404244
Encompass_Preferred
4-Hydroxyindole is a type of hydroxyindole in which the 1H-indole at position 4 is substituted by a hydroxyl group. It serves as an important raw material or intermediate in the synthesis of pharmaceutical products and industrial polymers. This compound inhibits amyloid fibrillization and induces liver function impairment, thyroid abnormalities, and blood glucose fluctuations in mice, holding potential for research in neurodegenerative diseases and metabolic disorders.
  • Inhibits amyloid fibrillization
  • Induces liver function impairment, thyroid abnormalities, and blood glucose fluctuations
  • Targeted in biochemical assay reagents
  • Targeted for amyloid-β research
  • Relevant for neurological and metabolic disease research
Catalog No. 50-004-04244 Supplier Medchemexpress LLC Supplier No. HY345965G
4-Hydroxyindole is a type of hydroxyindole in which the 1H-indole at position 4 is substituted by a hydroxyl group. It serves as an important raw material or intermediate in the synthesis of pharmaceutical products and industrial polymers. This compound inhibits amyloid fibrillization and induces liver function impairment, thyroid abnormalities, and blood glucose fluctuations in mice, holding potential for research in neurodegenerative diseases and metabolic disorders.
  • Inhibits amyloid fibrillization
  • Induces liver function impairment, thyroid abnormalities, and blood glucose fluctuations
  • Targeted in biochemical assay reagents
  • Targeted for amyloid-β research
  • Relevant for neurological and metabolic disease research