Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroxyindole-3-acetic acid | 56395-08-5 | MFCD01326488 | 99.1% | 191.18 | C10H9NO3 | 10 MG
SDP

Catalog No. 5000284884
Encompass_Preferred

4-Hydroxyindole-3-acetic acid (4HIAA) is a chemical metabolite of psilocybin and psilocin used as a research reagent and analytical reference standard. Supplied as a solid suitable for laboratory assays, it supports biochemical studies, method development, and analytical verification.

  • Metabolite of psilocybin and psilocin.
  • Chemical formula C10H9NO3; molecular weight 191.18.
  • CAS number 56395-08-5.
  • High purity: 99.1% (HPLC) as reported on the COA.
  • Appearance: off-white to light yellow solid.
  • Storage: 4°C under nitrogen; in solvent store at -80°C for 6 months or -20°C for 1 month.
  • Available pack sizes: 5 mg, 10 mg, 25 mg.

Catalog No. 50-002-84884 Supplier Medchemexpress LLC Supplier No. HYW50938110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Hydroxyindole-3-acetic acid (4HIAA) is a chemical metabolite of psilocybin and psilocin used as a research reagent and analytical reference standard. Supplied as a solid suitable for laboratory assays, it supports biochemical studies, method development, and analytical verification.

  • Metabolite of psilocybin and psilocin.
  • Chemical formula C10H9NO3; molecular weight 191.18.
  • CAS number 56395-08-5.
  • High purity: 99.1% (HPLC) as reported on the COA.
  • Appearance: off-white to light yellow solid.
  • Storage: 4°C under nitrogen; in solvent store at -80°C for 6 months or -20°C for 1 month.
  • Available pack sizes: 5 mg, 10 mg, 25 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.