Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroxyindole-3-acetic acid | 56395-08-5 | MFCD01326488 | 99.1% | 191.18 | C10H9NO3 | 5 MG
SDP

Catalog No. 5000284085
Encompass_Preferred

4-Hydroxyindole-3-acetic acid (4HIAA) is an indole metabolite of psilocybin and psilocin used as a research reference standard and analytical reagent for studies of tryptamine metabolism.

  • High purity: 99.1% (HPLC) suitable for analytical applications.
  • Off-white to light yellow solid.
  • Molecular formula C10H9NO3; molecular weight 191.18.
  • Available in small research quantities: 5 mg, 10 mg, 25 mg.
  • Storage: store at 4°C under nitrogen; in solvent, -80°C up to 6 months, -20°C up to 1 month.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-84085 Supplier Medchemexpress LLC Supplier No. HYW5093815MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Hydroxyindole-3-acetic acid (4HIAA) is an indole metabolite of psilocybin and psilocin used as a research reference standard and analytical reagent for studies of tryptamine metabolism.

  • High purity: 99.1% (HPLC) suitable for analytical applications.
  • Off-white to light yellow solid.
  • Molecular formula C10H9NO3; molecular weight 191.18.
  • Available in small research quantities: 5 mg, 10 mg, 25 mg.
  • Storage: store at 4°C under nitrogen; in solvent, -80°C up to 6 months, -20°C up to 1 month.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.