Learn More
Medchemexpress LLC 4-Hydroxyindole (Standard) | 2380-94-1 | 97% | 133.15 | 100 MG
SDP
Catalog No. 5000405049
Encompass_Preferred
4-Hydroxyindole (Standard) is the analytical standard of 4-Hydroxyindole. This product is intended for research and analytical applications. 4-Hydroxyindole is a member of the class of hydroxyindoles that is 1H-indole substituted by a hydroxy group at position 4. 4-Hydroxyindole is an important raw material or intermediate in the synthesis of pharmaceutical products and industrial polymers.
  • Intended for research and analytical applications.
  • Member of the class of hydroxyindoles, specifically 1H-indole substituted by a hydroxy group at position 4.
  • Important raw material or intermediate in the synthesis of pharmaceutical products and industrial polymers.
  • Analytical standard used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-05049 Supplier Medchemexpress LLC Supplier No. HY34596R100MG
4-Hydroxyindole (Standard) is the analytical standard of 4-Hydroxyindole. This product is intended for research and analytical applications. 4-Hydroxyindole is a member of the class of hydroxyindoles that is 1H-indole substituted by a hydroxy group at position 4. 4-Hydroxyindole is an important raw material or intermediate in the synthesis of pharmaceutical products and industrial polymers.
  • Intended for research and analytical applications.
  • Member of the class of hydroxyindoles, specifically 1H-indole substituted by a hydroxy group at position 4.
  • Important raw material or intermediate in the synthesis of pharmaceutical products and industrial polymers.
  • Analytical standard used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.