Learn More
Medchemexpress LLC HY-N6858 5mg Medchemexpress, 4-Hydroxyisoleucine CAS:781658-23-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN68585MG
Encompass_Preferred
Medchemexpress, HY-N6858 5mg 4-Hydroxyisoleucine CAS:781658-23-9 4-Hydroxyisoleucine (4-​Hydroxy-​L-​isoleucine) is an amino acid which can be extracted and purified from fenugreek seeds. 4-Hydroxyisoleucine (4-​Hydroxy-​L-​isoleucine) displays an insulinotropic activity of great interest. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0477