Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxyisophthalaldehyde 98.0+%, TCI America™
SDP

Catalog No. F03105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03105G 5 g
F03101G 1 g
F031025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F03105G Supplier TCI America Supplier No. F03105G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3328-70-9
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.13
MDL Number MFCD00003334
InChI Key FEUATHOQKVGPEK-UHFFFAOYSA-N
Synonym 2,4-Diformylphenol
PubChem CID 165106
IUPAC Name 4-hydroxybenzene-1,3-dicarbaldehyde
SMILES OC1=CC=C(C=O)C=C1C=O

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 5 g
Formula Weight 150.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxyisophthalaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.