Learn More
Medchemexpress LLC 4-Hydroxyisophthalic acid | 636-46-4 | 99.5% | C8H6O5 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY7663210MM/1ML
Encompass_Preferred
This compound activates antioxidant enzymes (such as catalase and superoxide dismutase) and scavenges free radicals, demonstrating antioxidant properties. It also activates AChE and BChE, which enhances neuronal function and improves neurobehavioral defects. Additionally, it can ameliorate cognitive defects and circadian rhythm disorders.
  • Activates antioxidant enzymes and scavenges free radicals.
  • Exhibits antioxidant properties.
  • Enhances neuronal function.
  • Improves neurobehavioral defects.
  • Ameliorates cognitive defects and circadian rhythm disorders.
Catalog No. 50-004-33663