Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Hydroxyisophthalic acid | 636-46-4 | 99.5% | C8H6O5 | 25 MG
SDP

Catalog No. 5000359265
Encompass_Preferred

4-Hydroxyisophthalic acid activates antioxidant enzymes, such as catalase (CAT) and superoxide dismutase (SOD), scavenges free radicals, and exhibits antioxidant property. It also activates AChE and BChE, enhancing neuronal function and improving Tau-induced neurobehavioral defects. Additionally, it improves cognitive defects and ameliorates circadian rhythm disorders in fruit flies.

  • Activates antioxidant enzymes (CAT and SOD)
  • Scavenges free radicals
  • Exhibits antioxidant properties
  • Enhances neuronal function
  • Improves Tau-induced neurobehavioral defects
  • Ameliorates circadian rhythm disorders
  • Molecular weight: 182.13
  • Appearance: Solid
  • Color: Off-white to pink
  • Structure classification: Ketones, aldehydes, acids
  • Derived from plants Apocynaceae Decalepis hamiltonii Wight & Arn.
  • For research use only

Catalog No. 50-003-59265 Supplier Medchemexpress LLC Supplier No. 25MGHY76632
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Hydroxyisophthalic acid activates antioxidant enzymes, such as catalase (CAT) and superoxide dismutase (SOD), scavenges free radicals, and exhibits antioxidant property. It also activates AChE and BChE, enhancing neuronal function and improving Tau-induced neurobehavioral defects. Additionally, it improves cognitive defects and ameliorates circadian rhythm disorders in fruit flies.

  • Activates antioxidant enzymes (CAT and SOD)
  • Scavenges free radicals
  • Exhibits antioxidant properties
  • Enhances neuronal function
  • Improves Tau-induced neurobehavioral defects
  • Ameliorates circadian rhythm disorders
  • Molecular weight: 182.13
  • Appearance: Solid
  • Color: Off-white to pink
  • Structure classification: Ketones, aldehydes, acids
  • Derived from plants Apocynaceae Decalepis hamiltonii Wight & Arn.
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.