Learn More
Medchemexpress LLC 4-Hydroxyisophthalic acid | 636-46-4 | 99.5% | C8H6O5 | 50 MG
SDP
Catalog No. 5000434176
Encompass_Preferred
4-Hydroxyisophthalic acid activates antioxidant enzymes (such as catalase CAT and superoxide dismutase SOD), scavenges free radicals, and exhibits antioxidant properties. It also activates AChE and BChE, enhances neuronal function, improves Tau-induced neurobehavioral defects, and ameliorates cognitive defects and circadian rhythm disorders in fruit flies.
  • Activates antioxidant enzymes (catalase and superoxide dismutase)
  • Scavenges free radicals
  • Exhibits antioxidant properties
  • Activates AChE and BChE
  • Enhances neuronal function
  • Improves Tau-induced neurobehavioral defects
  • Ameliorates cognitive defects
  • Ameliorates circadian rhythm disorders in fruit flies
Catalog No. 50-004-34176 Supplier Medchemexpress LLC Supplier No. HY7663250MG
4-Hydroxyisophthalic acid activates antioxidant enzymes (such as catalase CAT and superoxide dismutase SOD), scavenges free radicals, and exhibits antioxidant properties. It also activates AChE and BChE, enhances neuronal function, improves Tau-induced neurobehavioral defects, and ameliorates cognitive defects and circadian rhythm disorders in fruit flies.
  • Activates antioxidant enzymes (catalase and superoxide dismutase)
  • Scavenges free radicals
  • Exhibits antioxidant properties
  • Activates AChE and BChE
  • Enhances neuronal function
  • Improves Tau-induced neurobehavioral defects
  • Ameliorates cognitive defects
  • Ameliorates circadian rhythm disorders in fruit flies