Learn More
Medchemexpress LLC 4-Hydroxyisophthalic acid | 636-46-4 | 99.51% | C8H6O5 | 100 MG
SDP
Catalog No. 5000433727
Encompass_Preferred
4-Hydroxyisophthalic acid is a compound known for its antioxidant properties. It functions by activating key antioxidant enzymes such as catalase (CAT) and superoxide dismutase (SOD), and by scavenging free radicals. This compound also enhances neuronal function through the activation of AChE and BChE, which can lead to improvements in neurobehavioral defects induced by Tau protein and can ameliorate circadian rhythm disorders.
  • Activates antioxidant enzymes (catalase CAT and superoxide dismutase SOD)
  • Scavenges free radicals
  • Exhibits antioxidant properties
  • Activates AChE and BChE
  • Enhances neuronal function
  • Improves Tau-induced neurobehavioral defects
  • Ameliorates circadian rhythm disorders in fruit flies
Catalog No. 50-004-33727 Supplier Medchemexpress LLC Supplier No. HY76632100MG
4-Hydroxyisophthalic acid is a compound known for its antioxidant properties. It functions by activating key antioxidant enzymes such as catalase (CAT) and superoxide dismutase (SOD), and by scavenging free radicals. This compound also enhances neuronal function through the activation of AChE and BChE, which can lead to improvements in neurobehavioral defects induced by Tau protein and can ameliorate circadian rhythm disorders.
  • Activates antioxidant enzymes (catalase CAT and superoxide dismutase SOD)
  • Scavenges free radicals
  • Exhibits antioxidant properties
  • Activates AChE and BChE
  • Enhances neuronal function
  • Improves Tau-induced neurobehavioral defects
  • Ameliorates circadian rhythm disorders in fruit flies