Learn More
Medchemexpress LLC 4-hydroxyisophthalic acid | 636-46-4 | MFCD00010391 | 99.5% | 182.13 g/mol | C8H6O5 | 10 MG
SDP
Catalog No. 5000298628
Encompass_Preferred
4-Hydroxyisophthalic acid is a small-molecule antioxidant used in biological research to modulate cholinesterase activity and reduce oxidative stress. It has been applied in studies of neuronal function, Tau-related neurobehavioral defects, and circadian rhythm effects in model organisms.
  • Activates antioxidant enzymes such as catalase and superoxide dismutase
  • Scavenges free radicals to reduce oxidative damage
  • Modulates acetylcholinesterase and butyrylcholinesterase activity
  • Applicable to studies of neuronal function and Tau-related behavioral defects
  • Supplied as a solid with high purity for research use
Catalog No. 50-002-98628 Supplier Medchemexpress LLC Supplier No. HY7663210MG
4-Hydroxyisophthalic acid is a small-molecule antioxidant used in biological research to modulate cholinesterase activity and reduce oxidative stress. It has been applied in studies of neuronal function, Tau-related neurobehavioral defects, and circadian rhythm effects in model organisms.
  • Activates antioxidant enzymes such as catalase and superoxide dismutase
  • Scavenges free radicals to reduce oxidative damage
  • Modulates acetylcholinesterase and butyrylcholinesterase activity
  • Applicable to studies of neuronal function and Tau-related behavioral defects
  • Supplied as a solid with high purity for research use