Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxyisophthalic Acid 98%, Thermo Scientific™

Catalog No. AC263021000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC263021000 100 g
AC263020250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC263021000 Supplier Thermo Scientific Chemicals Supplier No. 263021000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-46-4
Molecular Formula C8H6O5
Molecular Weight (g/mol) 182.131
InChI Key BCEQKAQCUWUNML-UHFFFAOYSA-N
Synonym 4-hydroxyisophthalic acid, eupirina, 4-hydroxy-m-phthalic acid, isophthalic acid, 4-hydroxy, 4-hipa, 4-hydroxy-1,3-benzenedicarboxylic acid, unii-p87ff77o00, 1,3-benzenedicarboxylic acid, 4-hydroxy, 4-hydroxy isophthalic acid, 4-hydroxyisophthalate
PubChem CID 12490
IUPAC Name 4-hydroxybenzene-1,3-dicarboxylic acid
SMILES C1=CC(=C(C=C1C(=O)O)C(=O)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Linear Formula HOC6H3(CO2H)2
Quantity 100 g
Beilstein 10, 502
Merck Index 15, 4866
Formula Weight 182.13
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 4-Hydroxyisophthalic acid, 98%
Show More Show Less
EINECSNumber 211-258-0
RTECSNumber NT2544000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.