Learn More
Medchemexpress LLC HY-N2208 5mg Medchemexpress, 4-Hydroxylonchocarpin CAS:56083-03-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN22085MG
Encompass_Preferred
Medchemexpress, HY-N2208 5mg 4-Hydroxylonchocarpin CAS:56083-03-5 4-Hydroxylonchocarpin is a chalcone compound from an extract of Psoralea corylifolia. 4-Hydroxylonchocarpin increases phosphorylation of p38 MAPK, JNK and ERK. 4-Hydroxylonchocarpin has diverse pharmacological activities, including antibacterial, antifungal, anticancer, antireverse transcriptase, antitubercular, antimalarial, anti-inflammatory and antioxidant activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2878