Learn More
Medchemexpress LLC HY-W008772 5mg Medchemexpress, 4-Hydroxymephenytoin CAS:61837-65-8 Purity:>98%
SDP
Catalog No. 501934522
Encompass_Preferred
Medchemexpress, HY-W008772 5mg 4-Hydroxymephenytoin CAS:61837-65-8 4-Hydroxymephenytoin is a metabolism of an antiepileptic drug mephenytoin, which is used as a CYP2C19 substrate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4522 Supplier Medchemexpress LLC Supplier No. HYW0087725MG
Medchemexpress, HY-W008772 5mg 4-Hydroxymephenytoin CAS:61837-65-8 4-Hydroxymephenytoin is a metabolism of an antiepileptic drug mephenytoin, which is used as a CYP2C19 substrate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.