Learn More
Medchemexpress LLC 4-Hydroxymephenytoin | 61837-65-8 | 99.8% | 234.25 | 1 MG
SDP
Catalog No. 5000366342
Encompass_Preferred
4-Hydroxymephenytoin is a metabolite of the antiepileptic agent mephenytoin and serves as a CYP2C19 substrate. It is intended for research use only and is not sold to patients. It is also used for developing UltraPerformance LC (UPLC) tandem mass spectrometry assays to evaluate CYP probe drugs and their relevant metabolites in human urine or plasma samples.
  • Utilized as a cyp2c19 substrate
  • Suitable for laboratory centrifugation and storage
  • Used in developing uplc tandem mass spectrometry assays
Catalog No. 50-003-66342 Supplier Medchemexpress LLC Supplier No. HYW008772S1MG
Add to Cart
Add to Cart
4-Hydroxymephenytoin is a metabolite of the antiepileptic agent mephenytoin and serves as a CYP2C19 substrate. It is intended for research use only and is not sold to patients. It is also used for developing UltraPerformance LC (UPLC) tandem mass spectrometry assays to evaluate CYP probe drugs and their relevant metabolites in human urine or plasma samples.
  • Utilized as a cyp2c19 substrate
  • Suitable for laboratory centrifugation and storage
  • Used in developing uplc tandem mass spectrometry assays