Learn More
Medchemexpress LLC 4-Hydroxymephenytoin | 61837-65-8 | 99.8% | 234.25 | 1 ML
SDP
Catalog No. 5000452655
Encompass_Preferred
4-Hydroxymephenytoin is a metabolism of an antiepileptic agent mephenytoin, which is used as a CYP2C19 substrate. It is used for developing UltraPerformance LC (UPLC) tandem mass spectrometry assays for evaluating CYP probe drugs and their relevant metabolites in human urine or plasma samples.
  • Metabolism of an antiepileptic agent mephenytoin
  • Used as a CYP2C19 substrate
  • Used for developing UltraPerformance LC (UPLC) tandem mass spectrometry assays
  • Evaluates CYP probe drugs and their relevant metabolites
  • Suitable for human urine or plasma samples
Catalog No. 50-004-52655 Supplier Medchemexpress LLC Supplier No. HYW00877210MM/1ML
Add to Cart
Add to Cart
4-Hydroxymephenytoin is a metabolism of an antiepileptic agent mephenytoin, which is used as a CYP2C19 substrate. It is used for developing UltraPerformance LC (UPLC) tandem mass spectrometry assays for evaluating CYP probe drugs and their relevant metabolites in human urine or plasma samples.
  • Metabolism of an antiepileptic agent mephenytoin
  • Used as a CYP2C19 substrate
  • Used for developing UltraPerformance LC (UPLC) tandem mass spectrometry assays
  • Evaluates CYP probe drugs and their relevant metabolites
  • Suitable for human urine or plasma samples