Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(Hydroxymethyl)benzeneboronic acid | 59016-93-2 | 151.96 | 1 ML
SDP

Catalog No. 5000446663
Encompass_Preferred

4-(Hydroxymethyl)benzeneboronic acid is an important chemical intermediate used for the synthesis of other active compounds. It is also involved in the synthesis of tetrahedral boronate ester.

  • Chemical formula: C7H9BO3
  • Appearance: White to off-white solid
  • Soluble in DMSO (100 mg/mL, requires ultrasonic)
  • Storage as powder: 3 years at -20°C, 2 years at 4°C
  • Storage in solvent: 6 months at -80°C, 1 month at -20°C
  • For research use only

Catalog No. 50-004-46663 Supplier Medchemexpress LLC Supplier No. HYW00094910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-(Hydroxymethyl)benzeneboronic acid is an important chemical intermediate used for the synthesis of other active compounds. It is also involved in the synthesis of tetrahedral boronate ester.

  • Chemical formula: C7H9BO3
  • Appearance: White to off-white solid
  • Soluble in DMSO (100 mg/mL, requires ultrasonic)
  • Storage as powder: 3 years at -20°C, 2 years at 4°C
  • Storage in solvent: 6 months at -80°C, 1 month at -20°C
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.