Learn More
Medchemexpress LLC 4-(Hydroxymethyl)benzeneboronic acid | 59016-93-2 | 151.96 | 10 G
SDP
Catalog No. 5000446662
Encompass_Preferred
4-(Hydroxymethyl)benzeneboronic acid is an important intermediate that can be used for the synthesis of other active compounds. It is also involved in the synthesis of tetrahedral boronate ester.
  • Used as an important intermediate for synthesizing active compounds
  • Involved in the synthesis of tetrahedral boronate ester
  • Suitable for laboratory chemical applications
  • Stable under recommended storage conditions
  • Appears as a white to off-white solid
  • For research use only
Catalog No. 50-004-46662 Supplier Medchemexpress LLC Supplier No. HYW00094910G
4-(Hydroxymethyl)benzeneboronic acid is an important intermediate that can be used for the synthesis of other active compounds. It is also involved in the synthesis of tetrahedral boronate ester.
  • Used as an important intermediate for synthesizing active compounds
  • Involved in the synthesis of tetrahedral boronate ester
  • Suitable for laboratory chemical applications
  • Stable under recommended storage conditions
  • Appears as a white to off-white solid
  • For research use only