Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Hydroxymethyl)cyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. H125725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H125725G 25 g
H12575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H125725G Supplier TCI America Supplier No. H125725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 33893-85-5
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
InChI Key VGRZISGVNOKTQU-UHFFFAOYSA-N
PubChem CID 12892866
IUPAC Name 4-(hydroxymethyl)cyclohexan-1-ol
SMILES C1CC(CCC1CO)O

Specifications

Color Yellow
Boiling Point 122°C
Quantity 25 g
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Hydroxymethyl)cyclohexanol (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.