Learn More
Medchemexpress LLC 4-(Hydroxymethyl)phenylacetic acid (4HMPAA) | 73401-74-8 | 166.18 | 100 G
SDP
Catalog No. 5000445608
Encompass_Preferred
4-(Hydroxymethyl)phenylacetic acid can serve as the main body of insoluble polypeptide. It can react with DEPBT and 4-acetylaniline to form amide intermediates. This compound is a metabolite produced by the metabolism of flavonoids such as quercetin by colonic microbiota. 4-(Hydroxymethyl)phenylacetic acid significantly inhibits 6-MP oxidation and has no significant inhibition on xanthine oxidation.
  • Serves as main body of insoluble polypeptide
  • Reacts with DEPBT and 4-acetylaniline to form amide intermediates
  • Metabolite produced by colonic microbiota
  • Significantly inhibits 6-MP oxidation
Catalog No. 50-004-45608 Supplier Medchemexpress LLC Supplier No. HYW004128100G
4-(Hydroxymethyl)phenylacetic acid can serve as the main body of insoluble polypeptide. It can react with DEPBT and 4-acetylaniline to form amide intermediates. This compound is a metabolite produced by the metabolism of flavonoids such as quercetin by colonic microbiota. 4-(Hydroxymethyl)phenylacetic acid significantly inhibits 6-MP oxidation and has no significant inhibition on xanthine oxidation.
  • Serves as main body of insoluble polypeptide
  • Reacts with DEPBT and 4-acetylaniline to form amide intermediates
  • Metabolite produced by colonic microbiota
  • Significantly inhibits 6-MP oxidation