Learn More
Medchemexpress LLC 4-(Hydroxymethyl)phenylacetic acid | 73401-74-8 | 166.18 | 1 ML
SDP
Catalog No. 5000447957
Encompass_Preferred
4-(Hydroxymethyl)phenylacetic acid can serve as the main body of insoluble polypeptide. It can react with DEPBT and 4-acetylaniline to form amide intermediates. This compound is a metabolite produced by the metabolism of flavonoids such as quercetin by colonic microbiota. It significantly inhibits 6-MP oxidation and has no significant inhibition on xanthine oxidation.
  • Can serve as the main body of insoluble polypeptide.
  • Can react with DEPBT and 4-acetylaniline to form amide intermediates.
  • A metabolite produced by the metabolism of flavonoids such as quercetin by colonic microbiota.
  • Significantly inhibits 6-MP oxidation.
  • Has no significant inhibition on xanthine oxidation.
Catalog No. 50-004-47957 Supplier Medchemexpress LLC Supplier No. HYW00412810MM/1ML
4-(Hydroxymethyl)phenylacetic acid can serve as the main body of insoluble polypeptide. It can react with DEPBT and 4-acetylaniline to form amide intermediates. This compound is a metabolite produced by the metabolism of flavonoids such as quercetin by colonic microbiota. It significantly inhibits 6-MP oxidation and has no significant inhibition on xanthine oxidation.
  • Can serve as the main body of insoluble polypeptide.
  • Can react with DEPBT and 4-acetylaniline to form amide intermediates.
  • A metabolite produced by the metabolism of flavonoids such as quercetin by colonic microbiota.
  • Significantly inhibits 6-MP oxidation.
  • Has no significant inhibition on xanthine oxidation.