Learn More
Medchemexpress LLC 4-(Hydroxymethyl)phenylacetic acid | 73401-74-8 | 166.18 | 25 G
SDP
Catalog No. 5000449470
Encompass_Preferred
4-(Hydroxymethyl)phenylacetic acid (4HMPAA) is a chemical compound that can serve as the main body of insoluble polypeptide. It is also a metabolite produced by the metabolism of flavonoids such as quercetin by colonic microbiota and significantly inhibits 6-MP oxidation.
  • Can serve as the main body of insoluble polypeptide
  • Reacts with DEPBT and 4-acetylaniline to form amide intermediates
  • Metabolite produced by the metabolism of flavonoids by colonic microbiota
  • Significantly inhibits 6-MP oxidation
  • Solid appearance, white to light yellow
  • Melting/freezing point of 131-134°C
  • Soluble in DMSO (100 mg/mL)
Catalog No. 50-004-49470 Supplier Medchemexpress LLC Supplier No. 25GHYW004128
4-(Hydroxymethyl)phenylacetic acid (4HMPAA) is a chemical compound that can serve as the main body of insoluble polypeptide. It is also a metabolite produced by the metabolism of flavonoids such as quercetin by colonic microbiota and significantly inhibits 6-MP oxidation.
  • Can serve as the main body of insoluble polypeptide
  • Reacts with DEPBT and 4-acetylaniline to form amide intermediates
  • Metabolite produced by the metabolism of flavonoids by colonic microbiota
  • Significantly inhibits 6-MP oxidation
  • Solid appearance, white to light yellow
  • Melting/freezing point of 131-134°C
  • Soluble in DMSO (100 mg/mL)