Learn More
Medchemexpress LLC 4-(Hydroxymethyl)phenylacetic acid | 73401-74-8 | 166.18 | 5 G
SDP
Catalog No. 5000449211
Encompass_Preferred
4-(Hydroxymethyl)phenylacetic acid (4HMPAA) serves as a main body of insoluble polypeptide. It can react with DEPBT and 4-acetylaniline to form amide intermediates. This compound is a metabolite produced by the metabolism of flavonoids such as quercetin by colonic microbiota. It significantly inhibits 6-MP oxidation and has no significant inhibition on xanthine oxidation.
  • Serves as main body of insoluble polypeptide.
  • Reacts with DEPBT and 4-acetylaniline to form amide intermediates.
  • Metabolite produced by metabolism of flavonoids such as quercetin.
  • Significantly inhibits 6-MP oxidation.
Catalog No. 50-004-49211 Supplier Medchemexpress LLC Supplier No. HYW0041285G
4-(Hydroxymethyl)phenylacetic acid (4HMPAA) serves as a main body of insoluble polypeptide. It can react with DEPBT and 4-acetylaniline to form amide intermediates. This compound is a metabolite produced by the metabolism of flavonoids such as quercetin by colonic microbiota. It significantly inhibits 6-MP oxidation and has no significant inhibition on xanthine oxidation.
  • Serves as main body of insoluble polypeptide.
  • Reacts with DEPBT and 4-acetylaniline to form amide intermediates.
  • Metabolite produced by metabolism of flavonoids such as quercetin.
  • Significantly inhibits 6-MP oxidation.