Learn More
Medchemexpress LLC 4-(Hydroxymethyl)phenylacetic acid | 73401-74-8 | 166.18 | 50 G
SDP
Catalog No. 5000449151
Encompass_Preferred
4-(Hydroxymethyl)phenylacetic acid is a metabolite resulting from the metabolism of flavonoids like quercetin by colonic microbiota. It acts as a main component of insoluble polypeptides and participates in reactions forming amide intermediates. This compound notably inhibits 6-MP oxidation, showing no significant effect on xanthine oxidation.
  • Acts as main body of insoluble polypeptide
  • Reacts to form amide intermediates
  • Metabolite of flavonoids such as quercetin
  • Significantly inhibits 6-MP oxidation
Catalog No. 50-004-49151 Supplier Medchemexpress LLC Supplier No. HYW00412850G
4-(Hydroxymethyl)phenylacetic acid is a metabolite resulting from the metabolism of flavonoids like quercetin by colonic microbiota. It acts as a main component of insoluble polypeptides and participates in reactions forming amide intermediates. This compound notably inhibits 6-MP oxidation, showing no significant effect on xanthine oxidation.
  • Acts as main body of insoluble polypeptide
  • Reacts to form amide intermediates
  • Metabolite of flavonoids such as quercetin
  • Significantly inhibits 6-MP oxidation