Learn More
Selleck Chemical LLC 4-Hydroxynonenal-5mg
SDP
Catalog No. 502390075
Encompass_Preferred
4-Hydroxynonenal (4-HNE, HNE, 4-hydroxy Nonenal, 4-hydroxy-2-nonenal) is one of the major end products of lipid peroxidation and has been widely accepted as an inducer of oxidative stress.
Catalog No. 50-239-0075 Supplier Selleck Chemical LLC Supplier No. S97935MG
4-Hydroxynonenal (4-HNE, HNE, 4-hydroxy Nonenal, 4-hydroxy-2-nonenal) is one of the major end products of lipid peroxidation and has been widely accepted as an inducer of oxidative stress.