Learn More
Medchemexpress LLC 4 Hydroxynonenal Antibody | 100 UL
SDP
Catalog No. 5000442715
Encompass_Preferred
4 Hydroxynonenal Antibody is a Rabbit-derived, non-conjugated IgG polyclonal antibody designed for research use. It specifically targets 4 Hydroxynonenal, an α,β unsaturated hydroxyalkenal and an oxidative/nitrosative stress biomarker. The antibody is affinity purified and supplied as a liquid formulation for various applications.
  • Targets 4 Hydroxynonenal, an oxidative/nitrosative stress biomarker
  • Rabbit-derived polyclonal antibody
  • Affinity purified for high specificity
  • Species-independent reactivity
  • Suitable for Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), and Immunofluorescence-Tissue (IF-Tissue) applications
  • Formulated in 0.01M TBS (pH7.4) with 1% BSA, 0.03% Proclin300, and 50% Glycerol
Catalog No. 50-004-42715 Supplier Medchemexpress LLC Supplier No. HYP81208100UL
Add to Cart
Add to Cart
4 Hydroxynonenal Antibody is a Rabbit-derived, non-conjugated IgG polyclonal antibody designed for research use. It specifically targets 4 Hydroxynonenal, an α,β unsaturated hydroxyalkenal and an oxidative/nitrosative stress biomarker. The antibody is affinity purified and supplied as a liquid formulation for various applications.
  • Targets 4 Hydroxynonenal, an oxidative/nitrosative stress biomarker
  • Rabbit-derived polyclonal antibody
  • Affinity purified for high specificity
  • Species-independent reactivity
  • Suitable for Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), and Immunofluorescence-Tissue (IF-Tissue) applications
  • Formulated in 0.01M TBS (pH7.4) with 1% BSA, 0.03% Proclin300, and 50% Glycerol