Learn More
Abcam 4-Hydroxynonenal ELISA Kit, 96TEST
Supplier:  Abcam AB28780396TEST
Encompass_Preferred
4-Hydroxynonenal ELISA Kit ab287803 is used to quantify 4-Hydroxynonenal with a sensitivity of 5 pg/ml. Competitive ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5728