Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Hydroxyphenoxy)acetic acid, 98%, Thermo Scientific Chemicals

Catalog No. AC412090050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC412090050 5 g Glass bottle
AC412090250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC412090050 Supplier Thermo Scientific Chemicals Supplier No. 412090050
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1878-84-8
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00014362
InChI Key PKGWLCZTTHWKIZ-UHFFFAOYSA-N
Synonym 4-hydroxyphenoxyacetic acid, 4-hydroxyphenoxy acetic acid, 2-4-hydroxyphenoxy acetic acid, 4-hydroxypheoxyacetate, acetic acid, 4-hydroxyphenoxy, 4-hydroxy-phenoxy-acetic acid, p-hydroxyphenoxyacetic acid, acetic acid, 2-4-hydroxyphenoxy, acetic acid, p-hydroxyphenoxy, 4-carboxymethoxy phenol
PubChem CID 15881
ChEBI CHEBI:1881
IUPAC Name 2-(4-hydroxyphenoxy)acetic acid
SMILES C1=CC(=CC=C1O)OCC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 152°C to 156°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 168.15
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material (4-Hydroxyphenoxy)acetic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-525-8
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.