Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (4-Hydroxyphenyl)(4-(Prop-2-Yn-1-Yloxy)Phenyl)Methanone

Catalog No. 900615100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-061-5100MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-061-5100MG Supplier Sigma Aldrich Supplier No. 900615100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(4-Hydroxyphenyl)(4-(prop-2-yn-1-yloxy)phenyl)methanone is used for chemical probe synthesis, this trifunctional building block contains a light-activated benzophenone, alkyne tag, and hydroxyl synthetic handle. When appended to a ligand or pharmacophore through its hydroxyl linker, this building block allows for UV light-induced covalent modification of a biological target with potential for downstream applications via the alkyne tag. Use alone or in parallel with other multi-functional building blocks to discover the optimal probe for your chemical biology experiments.Technology Spotlight: Trifuctional Probe Building BlocksA library approach to rapidly discover photoaffinity probes of the mRNA decapping scavenger enzyme DcpSDesign, synthesis and biological evaluation of photoaffinity probes of antiangiogenic homoisoflavonoidsClickable Photoaffinity Ligands for Metabotropic Glutamate Receptor 5 Based on Select Acetylenic Negative Allosteric Modulatorsgamma-Secretase Modulator (GSM) Photoaffinity Probes Reveal Distinct Allosteric Binding Sites on Presenilin

Specifications

CAS 2140866-79-9
Molecular Formula C21H20O5
Linear Formula C21H20O5
Quantity 50 mg
Synonym Carboxyl benzophenone alkyne; Probe building block
Molecular Weight (g/mol) 352.38 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.