Learn More
Medchemexpress LLC 4-Hydroxyphenyl acetate | 3233-32-7 | 99.88% | 152.15 | 1 ML
SDP
Catalog No. 5000400434
Encompass_Preferred
4-Hydroxyphenyl acetate (4-HPA) is identified as a natural antioxidant that provides protection to cells against necrosis induced by oxidative stress. It functions by inhibiting the increase of cellular reactive oxygen species (ROS) resulting from oxidative stress. Furthermore, it enhances the expression of NQO1 and HO-1 genes through the stabilization and nuclear translocation of the NRF2 transcription factor.
  • Protects cells from oxidative stress-induced necrosis.
  • Blocks the increase of cellular ROS induced by oxidative stress.
  • Up-regulates NQO1 and HO-1 genes.
  • Stabilizes and induces nuclear translocation of NRF2 transcription factor.
Catalog No. 50-004-00434 Supplier Medchemexpress LLC Supplier No. HY3026710MM/1ML
4-Hydroxyphenyl acetate (4-HPA) is identified as a natural antioxidant that provides protection to cells against necrosis induced by oxidative stress. It functions by inhibiting the increase of cellular reactive oxygen species (ROS) resulting from oxidative stress. Furthermore, it enhances the expression of NQO1 and HO-1 genes through the stabilization and nuclear translocation of the NRF2 transcription factor.
  • Protects cells from oxidative stress-induced necrosis.
  • Blocks the increase of cellular ROS induced by oxidative stress.
  • Up-regulates NQO1 and HO-1 genes.
  • Stabilizes and induces nuclear translocation of NRF2 transcription factor.