Learn More
Medchemexpress LLC 4-hydroxyphenyl diphenyl phosphate | 56806-74-7 | MFCD30529787 | 342.28 | C18H15O5P | 50 MG
SDP
Catalog No. 5000281102
Encompass_Preferred
4-Hydroxyphenyl diphenyl phosphate is a hydroxylated metabolite of organophosphate flame retardants used as a research reference or standard in metabolism and exposure studies. The substance is characterized by molecular formula C18H15O5P and molecular weight 342.28, and is supplied with manufacturer documentation and storage guidance for research use only.
  • Hydroxylated metabolite of organophosphate flame retardants.
  • Intended for use as a research reference standard.
  • Molecular formula C18H15O5P and molecular weight 342.28.
  • Supplied with documentation and certificate of analysis guidance for storage.
  • For research use only; not for human therapeutic or clinical applications.
Catalog No. 50-002-81102 Supplier Medchemexpress LLC Supplier No. HYW39931150MG
4-Hydroxyphenyl diphenyl phosphate is a hydroxylated metabolite of organophosphate flame retardants used as a research reference or standard in metabolism and exposure studies. The substance is characterized by molecular formula C18H15O5P and molecular weight 342.28, and is supplied with manufacturer documentation and storage guidance for research use only.
  • Hydroxylated metabolite of organophosphate flame retardants.
  • Intended for use as a research reference standard.
  • Molecular formula C18H15O5P and molecular weight 342.28.
  • Supplied with documentation and certificate of analysis guidance for storage.
  • For research use only; not for human therapeutic or clinical applications.