Learn More
Medchemexpress LLC 4-hydroxyphenyl diphenyl phosphate | 56806-74-7 | MFCD30529787 | 342.28 g/mol | C18H15O5P | 25 MG
SDP
Catalog No. 5000281052
Encompass_Preferred
4-Hydroxyphenyl diphenyl phosphate is a hydroxylated metabolite of organophosphate flame retardants provided as a research chemical for studies of metabolism, environmental fate, and toxicology. It is used as an analytical reference and test compound in method development and quantitation of flame retardant metabolites.
  • Used as a metabolite reference in environmental and toxicology studies.
  • Suitable for analytical method development and quantitation.
  • Molecular formula C18H15O5P and molecular weight 342.28 g/mol.
  • Cataloged under CAS number 56806-74-7 for unambiguous identification.
  • Available in small research quantities (for example, 25 mg).
  • Intended for research use only; not for human or diagnostic use.
Catalog No. 50-002-81052 Supplier Medchemexpress LLC Supplier No. HYW39931125MG
4-Hydroxyphenyl diphenyl phosphate is a hydroxylated metabolite of organophosphate flame retardants provided as a research chemical for studies of metabolism, environmental fate, and toxicology. It is used as an analytical reference and test compound in method development and quantitation of flame retardant metabolites.
  • Used as a metabolite reference in environmental and toxicology studies.
  • Suitable for analytical method development and quantitation.
  • Molecular formula C18H15O5P and molecular weight 342.28 g/mol.
  • Cataloged under CAS number 56806-74-7 for unambiguous identification.
  • Available in small research quantities (for example, 25 mg).
  • Intended for research use only; not for human or diagnostic use.