Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroxyphenyl diphenyl phosphate | 56806-74-7 | MFCD30529787 | 95.0% | 342.28 g·mol⁻¹ | C18H15O5P | 100 MG
SDP

Catalog No. 5000281334
Encompass_Preferred

4-Hydroxyphenyl diphenyl phosphate is a hydroxylated metabolite of organophosphate flame retardants supplied for research use. It is intended for use as an analytical reference and in studies of metabolism, toxicology, and environmental fate, and is provided as a solid in multiple laboratory quantities.

  • Hydroxylated metabolite of organophosphate flame retardant.
  • Used as a reference standard in analytical chemistry and metabolism studies.
  • Suitable for environmental fate and toxicology research.
  • Provided as a solid in multiple laboratory quantities for convenient dosing.
  • Molecular formula C18H15O5P; molecular weight 342.28 g·mol⁻¹.
  • CAS number 56806-74-7.

Catalog No. 50-002-81334 Supplier Medchemexpress LLC Supplier No. HYW399311100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Hydroxyphenyl diphenyl phosphate is a hydroxylated metabolite of organophosphate flame retardants supplied for research use. It is intended for use as an analytical reference and in studies of metabolism, toxicology, and environmental fate, and is provided as a solid in multiple laboratory quantities.

  • Hydroxylated metabolite of organophosphate flame retardant.
  • Used as a reference standard in analytical chemistry and metabolism studies.
  • Suitable for environmental fate and toxicology research.
  • Provided as a solid in multiple laboratory quantities for convenient dosing.
  • Molecular formula C18H15O5P; molecular weight 342.28 g·mol⁻¹.
  • CAS number 56806-74-7.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.