Learn More
Medchemexpress LLC 4-hydroxyphenyl diphenyl phosphate | 56806-74-7 | MFCD30529787 | 95.0% | 342.28 g/mol | C18H15O5P | 5 MG
SDP
Catalog No. 5000281546
Encompass_Preferred
4-Hydroxyphenyl diphenyl phosphate is a hydroxylated metabolite of organophosphate flame retardants commonly used as a research reagent in environmental, toxicological, and analytical studies. The compound has formula C18H15O5P and a molecular weight of 342.28 g/mol. Suppliers commonly offer mg-scale packs for laboratory use; consult the certificate of analysis for lot-specific purity and storage conditions.
  • Hydroxylated metabolite of organophosphate flame retardants.
  • Suitable for environmental and toxicological studies.
  • Provided in small, research-oriented pack sizes (mg scale).
  • Formula C18H15O5P and molecular weight 342.28 g/mol.
  • Certificate of Analysis available for lot-specific purity and storage.
Catalog No. 50-002-81546 Supplier Medchemexpress LLC Supplier No. HYW3993115MG
4-Hydroxyphenyl diphenyl phosphate is a hydroxylated metabolite of organophosphate flame retardants commonly used as a research reagent in environmental, toxicological, and analytical studies. The compound has formula C18H15O5P and a molecular weight of 342.28 g/mol. Suppliers commonly offer mg-scale packs for laboratory use; consult the certificate of analysis for lot-specific purity and storage conditions.
  • Hydroxylated metabolite of organophosphate flame retardants.
  • Suitable for environmental and toxicological studies.
  • Provided in small, research-oriented pack sizes (mg scale).
  • Formula C18H15O5P and molecular weight 342.28 g/mol.
  • Certificate of Analysis available for lot-specific purity and storage.