Learn More
Medchemexpress LLC 4-hydroxyphenyl diphenyl phosphate | 56806-74-7 | MFCD30529787 | 95.0% | 342.3 g/mol | C18H15O5P | 10 MG
SDP
Catalog No. 5000281545
Encompass_Preferred
4-Hydroxyphenyl diphenyl phosphate is a hydroxylated metabolite of organophosphate flame retardants, supplied as a research reagent and analytical standard for studies of metabolism, environmental fate, and toxicology. It is commonly used in laboratory quantitation and exposure assessment workflows (CAS 56806-74-7, formula C18H15O5P).
  • Used as an analytical standard for LC-MS and GC-MS quantitation.
  • Serves as a reference material for metabolic and toxicological studies.
  • Applicable to environmental monitoring and exposure assessment.
  • Reported high purity to support reproducible experimental results.
Catalog No. 50-002-81545 Supplier Medchemexpress LLC Supplier No. HYW39931110MG
4-Hydroxyphenyl diphenyl phosphate is a hydroxylated metabolite of organophosphate flame retardants, supplied as a research reagent and analytical standard for studies of metabolism, environmental fate, and toxicology. It is commonly used in laboratory quantitation and exposure assessment workflows (CAS 56806-74-7, formula C18H15O5P).
  • Used as an analytical standard for LC-MS and GC-MS quantitation.
  • Serves as a reference material for metabolic and toxicological studies.
  • Applicable to environmental monitoring and exposure assessment.
  • Reported high purity to support reproducible experimental results.