Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Hydroxyphenyl)phosphonic Acid 97.0+%, TCI America™
SDP

Catalog No. H1559200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1559200MG 200 mg
H15591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1559200MG Supplier TCI America Supplier No. H1559200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 33795-18-5
Molecular Formula C6H7O4P
Molecular Weight (g/mol) 174.092
MDL Number MFCD01112120
InChI Key YIDVLWDHYNWHMH-UHFFFAOYSA-N
PubChem CID 214694
IUPAC Name (4-hydroxyphenyl)phosphonic acid
SMILES C1=CC(=CC=C1O)P(=O)(O)O

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 200 mg
Formula Weight 174.09
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (4-Hydroxyphenyl)phosphonic Acid
RTECSNumber SZ8566300
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.