Learn More
Medchemexpress LLC 4-Hydroxyphenylacetic acid | 156-38-7 | MFCD00004347 | 152.15 | 50 G
SDP
Catalog No. 5000415423
Encompass_Preferred
4-Hydroxyphenylacetic acid is a major microbiota-derived metabolite of polyphenols, known for its antioxidative action. It induces the expression of Nrf2, increasing Nrf2 translocation to the nucleus and enhancing the activity of phase II and antioxidant enzymes. This product is intended for research use only.
  • Increases Nrf2 translocation to the nucleus.
  • Enhances the activity of phase II and antioxidant enzymes.
  • Up-regulates UGT1A1, UGT1A9, and SULT2A1.
  • Suppresses CYP2E1 expression.
  • Functions as a human endogenous metabolite.
  • Functions as a microbial metabolite.
Catalog No. 50-004-15423 Supplier Medchemexpress LLC Supplier No. HYN190250G
Add to Cart
Add to Cart
4-Hydroxyphenylacetic acid is a major microbiota-derived metabolite of polyphenols, known for its antioxidative action. It induces the expression of Nrf2, increasing Nrf2 translocation to the nucleus and enhancing the activity of phase II and antioxidant enzymes. This product is intended for research use only.
  • Increases Nrf2 translocation to the nucleus.
  • Enhances the activity of phase II and antioxidant enzymes.
  • Up-regulates UGT1A1, UGT1A9, and SULT2A1.
  • Suppresses CYP2E1 expression.
  • Functions as a human endogenous metabolite.
  • Functions as a microbial metabolite.