Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxyphenylacetone 97.0+%, TCI America™
SDP

Catalog No. H124925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H124925G 25 g
H12495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H124925G Supplier TCI America Supplier No. H124925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 770-39-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00210439
InChI Key VWMVAQHMFFZQGD-UHFFFAOYSA-N
Synonym 4-hydroxyphenylacetone, 1-4-hydroxyphenyl propan-2-one, 1-4-hydroxyphenyl acetone, p-hydroxyphenylacetone, 1-4-hydroxyphenyl-2-propanone, 2-propanone, 1-4-hydroxyphenyl, 4-hydroxyphenyl acetone, 4-hydroxyphenylpropanone, acmc-209p7o, 4-hydroxyphenylpropan-2-one
PubChem CID 7019274
IUPAC Name 1-(4-hydroxyphenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(O)C=C1

Specifications

Melting Point 40°C
Color White-Yellow
Boiling Point 177°C
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Hydroxyphenylacetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.