Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. H12281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12281G 1 g
H1228-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12281G Supplier TCI America Supplier No. H12281G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material 4-Hydroxyphenylboronic Acid (contains varying amounts of Anhydride)
CAS 71597-85-8
Melting Point 232°C
Molecular Formula C6H7BO3
Quantity 1 g
MDL Number MFCD01074628
Synonym 4-hydroxyphenyl boronic acid, 4-hydroxybenzeneboronic acid, 4-boronophenol, 4-hydroxyphenyl boranediol, p-hydroxyphenylboronic acid, 4-hydroxyphenyl-boronic acid, boronic acid, 4-hydroxyphenyl, pubchem1729, acmc-209ojb
InChI Key COIQUVGFTILYGA-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(O)C=C1
IUPAC Name (4-hydroxyphenyl)boronic acid
Molecular Weight (g/mol) 137.93
PubChem CID 2734360
Formula Weight 137.93
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.