Learn More
Cayman Chemical 4-Hydroxyphenylpyruvic Acid, Purity: ≥95%, CAS: 156-39-8, 1 g
SDP
Catalog No. 502916798
Encompass_Preferred
An intermediate in aromatic amino acid metabolism; inhibits LPS-induced IL-6 and NO production in RAW 264.7 macrophages (IC50s = 2 and 0.5 mM, respectively); decreases serum starvation-induced mitochondrial superoxide production in EA.hy926 endothelial cells in vitro; increases survival in a rat model of hemorrhagic shock; serum and urinary levels are increased in patients with alkaptonuria
Catalog No. 50-291-6798 Supplier Cayman Chemical Supplier No. 385081 G
An intermediate in aromatic amino acid metabolism; inhibits LPS-induced IL-6 and NO production in RAW 264.7 macrophages (IC50s = 2 and 0.5 mM, respectively); decreases serum starvation-induced mitochondrial superoxide production in EA.hy926 endothelial cells in vitro; increases survival in a rat model of hemorrhagic shock; serum and urinary levels are increased in patients with alkaptonuria