Learn More
AdipoGen 4-Hydroxyphenylpyruvic acid
SDP
Catalog No. 502560803
Encompass_Preferred
Chemical. CAS 156-39-8. Formula C9H8O4. MW 180.16. Synthetic Keto acid that is involved in the tyrosine catabolism pathway. It is a product of the enzyme R-4-hydroxyphenyllactate dehydrogenase and is formed during tyrosine metabolism. The conversion from tyrosine to 4-HPPA is catalyzed by tyrosine aminotransferase. 4-HPPA can be converted to homogentisic acid which is one of the precursors to ochronotic pigment. A deficiency in the catalytic activity of HPD is known to lead to tyrosinemia type III, an autosomal recessive disorder characterized by elevated levels of blood tyrosine and massive excretion of tyrosine derivatives into urine.
Catalog No. 50-256-0803 Supplier AdipoGen Supplier No. CDXH0013G005
Chemical. CAS 156-39-8. Formula C9H8O4. MW 180.16. Synthetic Keto acid that is involved in the tyrosine catabolism pathway. It is a product of the enzyme R-4-hydroxyphenyllactate dehydrogenase and is formed during tyrosine metabolism. The conversion from tyrosine to 4-HPPA is catalyzed by tyrosine aminotransferase. 4-HPPA can be converted to homogentisic acid which is one of the precursors to ochronotic pigment. A deficiency in the catalytic activity of HPD is known to lead to tyrosinemia type III, an autosomal recessive disorder characterized by elevated levels of blood tyrosine and massive excretion of tyrosine derivatives into urine.