Learn More
Enzo Life Sciences 4-Hydroxyphenylretinamide (5mg). CAS: 65646-68-6
SDP
Supplier:  Enzo Life Sciences BMLGR1030005
Encompass_Preferred
An analog of all trans retinoic acid that shows antiproliferative activity on cultured human breast cancer cells. Highly selective activator of retinoid receptors. Induces apoptosis in malignant hemopoietic cell lines. Alternative name: Fenretinide. Purity: ≥97% (TLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0915