Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Hydroxypiperidine

Catalog No. 12877525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-877-525G 25 g
12-877-5100G 100 g
12-877-55G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-877-525G Supplier Sigma Aldrich Supplier No. 12877525G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The molecular structure, vibrational spectra, nbo and UV-spectral analysis of 4-hydroxypiperidine have been studied
  • The compounds with a substituted 4-piperidinol core have been found to be potent antagonists of the human h(3) receptor.4-hydroxypiperidine (4-piperidinol) can be used in the synthesis of a highly potent and selective ip (pgi(2) receptor) agonist
  • It can also be used in the study of copper-catalyzed n- versus o-arylation

Specifications

CAS 5382-16-1
Boiling Point 108°C to 114°C (10 mmHg)
Molecular Formula C5H11NO
Linear Formula C5H11NO
MDL Number MFCD00005999
Quantity 25 g
Synonym 4-Piperidinol
Molecular Weight (g/mol) 101.15
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.