Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxypiperidine, Contains Varying Amounts of Moisture 97%, Thermo Scientific™

Catalog No. AC156421000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC156421000 100 g
AC156420250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC156421000 Supplier Thermo Scientific Chemicals Supplier No. 156421000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5382-16-1
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
InChI Key HDOWRFHMPULYOA-UHFFFAOYSA-N
Synonym 4-hydroxypiperidine, 4-piperidinol, 4-hydroxy piperidine, 4-hydroxypiperidin, piperidine-4-ol, piperdine-4-ol, piperid-4-ol, 4-hydoxypiperidine, 4-hydroxypiperdine, 4-hyroxypiperidine
PubChem CID 79341
IUPAC Name piperidin-4-ol
SMILES C1CNCCC1O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Orange-Yellow to Yellow
Infrared Spectrum Authentic
Quantity 100 g
Beilstein 21, I, 188
Formula Weight 101.15
Percent Purity 96.0 to 102.0% Total base (on dry substance), ≥99.0% (by GC)
Physical Form Wet Solid
Chemical Name or Material 4-Hydroxypiperidine, 97%
Hazard Category HYGROSCOPIC
EINECSNumber 226-373-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.