Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxypyridine 99.0+%, TCI America™
SDP

Catalog No. H033225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H033225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H033225G Supplier TCI America Supplier No. H033225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory reagent with min. 99.0% purity (GC) used for chemical applications.

Chemical Identifiers

CAS 626-64-2
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.10
MDL Number MFCD00006419,MFCD00040458
InChI Key GCNTZFIIOFTKIY-UHFFFAOYSA-N
Synonym 4-hydroxypyridine, pyridin-4-ol, 4-pyridinol, 4-pyridone, 4 1h-pyridinone, pyridin-4 1h-one, 4-oxopyridine, 4-pyridinone, 4-pyridol, gamma-hydroxypyridine
PubChem CID 12290
IUPAC Name 1,4-dihydropyridin-4-one
SMILES O=C1C=CNC=C1

Specifications

Melting Point 148°C
Boiling Point 235°C
Quantity 25 g
Formula Weight 95.10
Percent Purity ≥99.0% (GC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxypyridine
EINECSNumber (5)-0722
RTECSNumber UU7701450
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.