Learn More
Cayman Chemical 4-HydroxyquInolIn 25g
SDP
Catalog No. 502910473
Encompass_Preferred
A quinoline alkaloid; is a core component of alkyl-4-hydroxyquinoline quorum sensing molecules; has been used as a precursor in the synthesis of compounds with antibacterial activity; has been used as a fluorescent detection reagent for iron(III) in bovine liver samples
Catalog No. 50-291-0473 Supplier Cayman Chemical Supplier No. 3506425 G
A quinoline alkaloid; is a core component of alkyl-4-hydroxyquinoline quorum sensing molecules; has been used as a precursor in the synthesis of compounds with antibacterial activity; has been used as a fluorescent detection reagent for iron(III) in bovine liver samples