Learn More
Medchemexpress LLC HY-16950 10mg , 4-Hydroxytamoxifen CAS:68047-06-3 Purity:>98%
SDP
Catalog No. 502259111
Encompass_Preferred
HY-16950 10mg 4-Hydroxytamoxifen CAS: 68047-06-3 4-Hydroxytamoxifen ((Z)-4-Hydroxytamoxifen) is an orally active, selective estrogen receptor modulator (SERM). 4-Hydroxytamoxifen ((Z)-4-Hydroxytamoxifen) induces CRISPR/Cas9 systems based on ER mediated nucleus translocation[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9111 Supplier Medchemexpress LLC Supplier No. HY1695010MG
HY-16950 10mg 4-Hydroxytamoxifen CAS: 68047-06-3 4-Hydroxytamoxifen ((Z)-4-Hydroxytamoxifen) is an orally active, selective estrogen receptor modulator (SERM). 4-Hydroxytamoxifen ((Z)-4-Hydroxytamoxifen) induces CRISPR/Cas9 systems based on ER mediated nucleus translocation[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.