Learn More
Selleck Chemical LLC 4-Hydroxytamoxifen(Afimoxifene) 10mg 68392-35-8 Afimoxifene
SDP
Supplier:  Selleck Chemical LLC S782710MG
Encompass_Preferred
4-Hydroxytamoxifen (Afimoxifene) is the active metabolite of tamoxifen and a selective estrogen receptor (ER) modulator that is widely used in the therapeutic and chemopreventive treatment of breast cancer. It activates on-target modification frequency for intein-Cas9 variants. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5305